MilliporeSigma
US EPA Method 8082: GC Analysis of Aroclor 1016/1260 (PCBs) on SLB®-5ms
US EPA Method 8082: GC Analysis of Aroclor 1016/1260 (PCBs) on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SPB®-608 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SPB®-608 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 0.50
Equity-1701 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
Equity-1701 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.00 μm
Equity-1701 Capillary GC Column
L × I.D. 30 m × 0.53 mm, df 0.50 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.00 μm
SLB®-5ms Capillary GC Column
L × I.D. 15 m × 0.10 mm, df 0.10 μm
accessory
Product No.
Description
Pricing
Wheaton wide-mouth bottle
amber soda-lime glass bottle, capacity 1000 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 12 ea
guard column
Product No.
Description
Pricing
Non-Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.10 mm
Non-Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.25 mm
Non-Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.53 mm
recommended
Product No.
Description
Pricing
Polyimide Sealing Resin
pkg of 5 g
required but not provided
Product No.
Description
Pricing
"Y" GlasSeal Connector, Borosilicate Glass
pkg of 1 ea
"Y" GlasSeal Connector, Fused Silica
pkg of 1 ea
"Y" GlasSeal Connector, Fused Silica
pkg of 3 ea
standard
Product No.
Description
Pricing
Aroclor 1232 solution
certified reference material, 1000 μg/mL in isooctane
Aroclor 1242 solution
certified reference material, 1000 μg/mL in isooctane
Aroclor 1248 solution
certified reference material, 1000 μg/mL in isooctane
Aroclor 1254 solution
certified reference material, 1000 μg/mL in isooctane
Aroclor 1260 solution
certified reference material, 1000 μg/mL in isooctane
Aroclor 1262 solution
certified reference material, 1000 μg/mL in isooctane
Aroclor 1221 solution
certified reference material, 1000 μg/mL in isooctane
Aroclor 1254 solution
certified reference material, 200 μg/mL in methanol
Aroclor Standard Mix 1
certified reference material, in acetone: methanol (2:3) (varied)
Aroclor 1016 solution
certified reference material, 1000 μg/mL in isooctane
Pesticide Surrogate Spike Mix
certified reference material, TraceCERT®, 200 μg/mL each component in acetone, ampule of 1 mL
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
100 °C (2 min), 15 °C/min to 330 °C (3 min)
inj. temp.
250 °C
detector
ECD, 330 °C
carrier gas
helium, 25 cm/sec, constant
injection
2 μL splitless (0.75 min)
liner
4 mm I.D., single taper
sample
Aroclor standard mix 1 (46846-U) diluted to 500 ppb / 50 ppb (Aroclors / surrogates) in n-hexane
Description
Analysis Note
Aroclors are commercial mixtures of PCB congeners. Due to their stability, they ave been used in many different industrial and commercial applications. Because of their toxicity and ability to bioaccumulate, production of these materials ceased in 1977. This application demonstrates the separation of two common Aroclor mixtures, Aroclor 1016 and Aroclor 1260 on the SLB-5ms.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany