MilliporeSigma
GC Analysis of Waxes in Olive Oil on MET-Biodiesel
GC Analysis of Waxes in Olive Oil on MET-Biodiesel suitable for GC
CONDITIONS
column
MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 μm with integrated 2 m x 0.53 mm I.D. guard (28668-U)
oven
200 °C, 20 °C/min to 350 °C (15 min)
detector
FID, 380 °C
carrier gas
helium, 10 mL/min
injection
1 μL, cool on-column
sample
olive oil waxes