MilliporeSigma
GC Analysis of ‘Positive List′ Chemicals (Mix 5) on SLB®-5ms
GC Analysis of ‘Positive List’ Chemicals (Mix 5) on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
80 °C (2 min), 5 °C/min to 300 °C (4 min)
inj. temp.
250 °C
MSD interface
300 °C
scan range
m/z 40-450
carrier gas
helium, 1 mL/min constant
injection
1.0 μL splitless (1 min)
sample
each pesticide at 20 ppm in acetone
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany