MilliporeSigma
GC Analysis of FAMEs in Atlantic Salmon on Omegawax
GC Analysis of FAMEs in Atlantic Salmon on Omegawax suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Omegawax Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
Omegawax, 30 m x 0.25 mm I.D., 0.25μm (24136)
oven
170 °C, 1 °C/min to 225°C
inj. temp.
250 °C
detector
FID, 260 °C
carrier gas
helium, 1.2 mL/min
injection
1 μL, 100:1 split
liner
4 mm ID FocusLiner with taper
sample
1. Fish Oil from capsule, prepared according to AOAC Method 991.39 2. Atlantic, farm-raised salmon, prepared according to AOCS Official Method Ce 1k-09
Description
Analysis Note
Samples of farm-raised Atlantic salmon were prepared and analyzed for fatty acids. The salmon was extracted using an acid digestion /alkali hydrolysis followed by methylation as described in AOCS Official Method Ce 1k-09 (2). The resulting extracts were then analyzed by GC/FID on an Omegawax 250 capillary column as described in AOCS Official Method Ce 1i-07.
Legal Information
null