MilliporeSigma
ASTM D5134: GC Analysis of Hydrocarbons on Petrocol® DH 50.2
ASTM D5134: GC Analysis of Hydrocarbons on Petrocol® DH 50.2 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Petrocol® DH 50.2 Capillary GC Column
L × I.D. 50 m × 0.20 mm, df 0.50 μm
CONDITIONS
column
Petrocol DH 50.2, 50 m × 0.20 mm I.D., 0.50 μm (24133-U)
oven
35 °C (30 min), 2 °C/min to 200 °C (15 min)
inj. temp.
200 °C
detector
FID, 250 °C
carrier gas
helium, 19-21 cm/sec @ 35°C (see ASTM D5134)
injection
0.2 μL, 200:1 split
Description
Legal Information
Petrocol is a registered trademark of Merck KGaA, Darmstadt, Germany