MilliporeSigma
  • Resistance to two organophosphorus insecticides in New Zealand populations of the Australian sheep blowfly, Lucilia cuprina.
Resistance to two organophosphorus insecticides in New Zealand populations of the Australian sheep blowfly, Lucilia cuprina.
Medical and veterinary entomology (1994-07-01)
J A Wilson, A C Heath
ABSTRACT
A treated-surface test was used in a bioassay to detect insecticide resistance to the organophosphorus compounds diazinon and propetamphos, in larvae of field strains of Lucilia cuprina in New Zealand during 1990/91 and 1991/92 (December-April). Mean LC50s for both diazinon and propetamphos showed a significant increase between seasons although this is not necessarily indicative of a trend. Mean LC50s for diazinon increased from 1.16 to 1.60 mg/l (P < 0.001) and for propetamphos from 0.49 to 0.59 mg/l (P < 0.005) in pooled populations. Diazinon and propetamphos resistance was significantly correlated in both seasons, suggesting side-resistance between the two insecticides.
Product Description
Supelco
Propetamphos, PESTANAL®, analytical standard