MilliporeSigma
  • Zinc alleviates pain through high-affinity binding to the NMDA receptor NR2A subunit.
Zinc alleviates pain through high-affinity binding to the NMDA receptor NR2A subunit.
Nature neuroscience (2011-07-05)
Chihiro Nozaki, Angela Maria Vergnano, Dominique Filliol, Abdel-Mouttalib Ouagazzal, Anne Le Goff, Stéphanie Carvalho, David Reiss, Claire Gaveriaux-Ruff, Jacques Neyton, Pierre Paoletti, Brigitte L Kieffer
ABSTRACT
Zinc is abundant in the central nervous system and regulates pain, but the underlying mechanisms are unknown. In vitro studies have shown that extracellular zinc modulates a plethora of signaling membrane proteins, including NMDA receptors containing the NR2A subunit, which display exquisite zinc sensitivity. We created NR2A-H128S knock-in mice to investigate whether Zn2+-NR2A interaction influences pain control. In these mice, high-affinity (nanomolar) zinc inhibition of NMDA currents was lost in the hippocampus and spinal cord. Knock-in mice showed hypersensitivity to radiant heat and capsaicin, and developed enhanced allodynia in inflammatory and neuropathic pain models. Furthermore, zinc-induced analgesia was completely abolished under both acute and chronic pain conditions. Our data establish that zinc is an endogenous modulator of excitatory neurotransmission in vivo and identify a new mechanism in pain processing that relies on NR2A NMDA receptors. The study also potentially provides a molecular basis for the pain-relieving effects of dietary zinc supplementation.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-NMDAR1 Antibody, clone 54.1, clone 54.1, Chemicon®, from mouse
Sigma-Aldrich
Anti-NR2A Antibody, clone A12W, rabbit monoclonal, culture supernatant, clone A12W, Upstate®