Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900895

Sigma-Aldrich

Yeast Nitrogen Base Without Amino Acids

Vetec, reagent grade, powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500

grade

reagent grade

product line

Vetec

form

powder

pH

5.2-5.6(0.67% solution)

storage temp.

room temp

General description

Yeast Nitrogen Base is a highly-referenced growth medium used for the cultivation of yeast. This nutrient-rich microbial broth contains nitrogen, vitamins, trace elements and salts.

Components

Approximate Ingredients
KH2PO4, 1 g/L
(NH4)2SO4 5 g/L
MgSO4, 0.5 g/L
NaCl, 0.1 g/L
CaCl2, 0.1 g/L
inositol, 2 mg/L

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service