Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900558

Sigma-Aldrich

Isopropyl myristate

Vetec, reagent grade, 98%

Synonym(s):

Isopropyl tetradecanoate, Myristic acid isopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOCH(CH3)2
CAS Number:
Molecular Weight:
270.45
Beilstein:
1781127
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

refractive index

n20/D 1.434 (lit.)

bp

193 °C/20 mmHg (lit.)

mp

~3 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCC(=O)OC(C)C

InChI

1S/C17H34O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-17(18)19-16(2)3/h16H,4-15H2,1-3H3

InChI key

AXISYYRBXTVTFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shishu Goindi et al.
International journal of pharmaceutics, 495(2), 913-923 (2015-10-13)
In this paper, we report an ionic liquid-in-water (IL/w) microemulsion (ME) formulation which is able to solubilize etodolac (ETO), a poorly water soluble drug for topical delivery using BMIMPF6 (1-butyl-3-methylimidazolium hexafluorophosphate) as IL, Tween 80 as surfactant and ethanol as
Ji Zhang et al.
International journal of pharmaceutics, 421(1), 34-44 (2011-10-01)
In this study, microemulsion microstructures, key formulation variables, and their relationship to drug transdermal permeation enhancement were investigated. A microemulsion system with high water soluble capacity was developed, using isopropyl myristate, Labrasol, and Cremophor EL as oil, surfactant, and co-surfactant
Qingping Tian et al.
International journal of pharmaceutics, 426(1-2), 202-210 (2012-01-26)
To improve the skin permeation of naproxen with larger dosage, microemulsion with high content of naproxen was investigated for transdermal delivery and its solubilization mechanism was studied. Naproxen micoremulsions composed of 4% isopropyl myristate, 18% Tween 80, 18% ethanol and
T N Engelbrecht et al.
Skin pharmacology and physiology, 25(4), 200-207 (2012-05-23)
In order to elucidate the mode of action of the lipophilic penetration enhancer isopropyl myristate (IPM) on a molecular scale, we investigated oriented quaternary stratum corneum (SC) lipid model membranes based on ceramide AP, cholesterol, palmitic acid and cholesterol sulfate
Qian Zhang et al.
Pharmaceutical research, 30(1), 32-40 (2012-08-28)
In principle, maximum transepidermal fluxes of solutes should be similar for different vehicles, except when the solute or vehicle modifies the skin. Here we estimated maximum flux, stratum corneum solubility, diffusivity and permeability coefficient for a range of similarly sized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service