Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900352

Sigma-Aldrich

Lithium dodecyl sulfate

Vetec, reagent grade, ≥98.5%

Synonym(s):

Dodecyl lithium sulfate, Dodecyl sulfate lithium salt, Lauryl sulfate lithium salt, Lithium lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Li
CAS Number:
Molecular Weight:
272.33
Beilstein:
5186718
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

grade

reagent grade

description

anionic detergent

product line

Vetec

Assay

≥98.5%

mol wt

272.33 g/mol

CMC

7-10 mM (20-25°C)

SMILES string

[Li+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Li/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

YFVGRULMIQXYNE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Anionic detergent that may be used in place of SDS for electrophoresis in cold conditions.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric J Montemayor et al.
Nucleic acids research, 42(16), 10845-10855 (2014-08-16)
The enzymatic processing of cellular RNA molecules requires selective recognition of unique chemical and topological features. The unusual 2',5'-phosphodiester linkages in RNA lariats produced by the spliceosome must be hydrolyzed by the intron debranching enzyme (Dbr1) before they can be
Mohamed Bouraada et al.
Journal of hazardous materials, 153(3), 911-918 (2007-11-13)
Dodecylsulfate- and dodecyl benzene sulfonate-hydrotalcites were prepared by calcination-rehydratation method. The surfactants intercalation in the interlayer space of hydrotalcite were checked by PXRD and FTIR spectroscopy where the resulting materials were found to be similar to those reported in the
F Bruna et al.
Journal of hazardous materials, 225-226, 74-80 (2012-05-23)
Polycyclic aromatic hydrocarbons (PAHs) are considered as priority pollutants because of their high risk to human health. In this paper, we addressed the issue of using hydrotalcite-based nanocomposites as adsorbents of six low molecular weight PAHs (acenaphthene, fluorene, phenanthrene, anthracene
Anna Wahlström et al.
The FEBS journal, 275(20), 5117-5128 (2008-09-13)
The amyloid beta peptide (Abeta) with 39-42 residues is the major component of amyloid plaques found in brains of Alzheimer's disease patients, and soluble oligomeric peptide aggregates mediate toxic effects on neurons. The Abeta aggregation involves a conformational change of
Shintaro Ida et al.
Journal of the American Chemical Society, 130(43), 14038-14039 (2008-10-07)
One-nanometer-thick nickel hydroxide nanosheets were prepared by exfoliation of layered nickel hydroxides intercalated with dodecyl sulfate (DS) ions. The shape of the nanosheets was hexagonal, as was that of the layered nickel hydroxides intercalated with DS ions. The nickel hydroxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service