Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900307

Sigma-Aldrich

CAPS

Vetec, reagent grade, ≥98%

Synonym(s):

3-(Cyclohexylamino)-1-propanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11NH(CH2)3SO3H
CAS Number:
Molecular Weight:
221.32
Beilstein:
2835588
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥98%

form

powder

pH

3.0-7.0

pKa 

10.4

mp

>300 °C (lit.)

solubility

water: 0.176 g/mL, clear, colorless

SMILES string

OS(=O)(=O)CCCNC1CCCCC1

InChI

1S/C9H19NO3S/c11-14(12,13)8-4-7-10-9-5-2-1-3-6-9/h9-10H,1-8H2,(H,11,12,13)

InChI key

PJWWRFATQTVXHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tobias Reinhardt et al.
Molecules (Basel, Switzerland), 25(21) (2020-11-14)
Phosphonates are widely used as antiscalants for softening processes in drinking water treatment. To prevent eutrophication and accumulation in the sediment, it is desirable to remove them from the membrane concentrate before they are discharged into receiving water bodies. This
Frances E Lock et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(34), E3534-E3543 (2014-08-13)
Remnants of ancient transposable elements (TEs) are abundant in mammalian genomes. These sequences harbor multiple regulatory motifs and hence are capable of influencing expression of host genes. In response to environmental changes, TEs are known to be released from epigenetic
Daniele de Sanctis et al.
The Journal of biological chemistry, 280(29), 27222-27229 (2005-05-27)
Hemoglobins at high concentration have been isolated long ago from some insect larvae living in hypoxic environments. Conversely, a monomeric hemoglobin has been discovered recently in the fruit fly Drosophila melanogaster as intracellular protein expressed both in larvae and in
Jelena Zaitseva et al.
Acta crystallographica. Section D, Biological crystallography, 60(Pt 6), 1076-1084 (2004-05-26)
Nucleotide-binding domains (NBDs), which are roughly 27 kDa in size, are conserved components of the large family of ABC (ATP-binding cassette) transporters, which includes importers and exporters. NBDs, or ABC-ATPases, supply energy for the translocation of a vast range of
Adrian Sculptoreanu et al.
Experimental neurology, 219(2), 562-573 (2009-07-28)
Neurokinins (NK) released from terminals of dorsal root ganglion (DRG) neurons may control firing of these neurons by an autofeedback mechanism. In this study we used patch clamp recording techniques to determine if NKs alter excitability of rat L4-S3 DRG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service