Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900257

Sigma-Aldrich

Triethanolamine

Vetec, reagent grade, 97%

Synonym(s):

2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)3N
CAS Number:
Molecular Weight:
149.19
Beilstein:
1699263
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
reagent grade
Assay:
97%
technique(s):
hybridization: suitable
bp:
190-193 °C/5 mmHg (lit.)
vapor pressure:
0.01 mmHg ( 20 °C)

grade

reagent grade

vapor density

5.14 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

product line

Vetec

Assay

97%

form

viscous liquid

autoignition temp.

600 °F

expl. lim.

8.5 %

technique(s)

hybridization: suitable

refractive index

n20/D 1.485 (lit.)

pH

10.5-11.5 (25 °C, 149 g/L)

useful pH range

7.3-8.3

pKa (25 °C)

7.8

bp

190-193 °C/5 mmHg (lit.)

mp

17.9-21 °C (lit.)

density

1.124 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

OCCN(CCO)CCO

InChI

1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2

InChI key

GSEJCLTVZPLZKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethanolamine, a derivative of ammonia, belongs to the family of ethanolamine. It exhibits the properties of both amines and alcohols. Hence, it can react with acids to form salts and also undergo esterification. It is commonly used in the preparation of detergent raw materials, emulsifiers and in cement production, etc. It is also used as a chemical absorbent to remove acid gases like CO2 and H2S.

Application

Triethanolamine can be used:
  • As an electron donor in H2 evolution studies under photocatalytic conditions.
  • As a template in the synthesis of a mesoporous catalyst comprising iron (Fe-TUD-1), which is used in Friedel-Crafts benzylation.
  • As a template and a metal complexing agent in the synthesis of a mesoporous catalyst comprising zirconium (Zr-TUD-1), which is used in Meerwein-Ponndorf-Verley reduction.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

354.2 °F - closed cup

Flash Point(C)

179 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zr-TUD-1: A novel heterogeneous catalyst for the Meerwein-Ponndorf-Verley reaction
Ramanathan A, et al.
J. Mol. Catal. A: Chem., 260(1-2), 62-69 (2006)
Chemical absorption of carbon dioxide with triethanolamine in non-aqueous solutions
Park S, et al.
Korean Journal of Chemical Engineering, 23(1), 138-143 (2006)
Ethanolamines and Propanolamines
Ullmann's Encyclopedia of Industrial Chemistry (2001)
One-step construction of FeOx modified g-C3N4 for largely enhanced visible-light photocatalytic hydrogen evolution
Cheng R, et al.
Carbon, 101(1), 62-70 (2016)
Facile construction of CuFe2 O 4/gC 3 N 4 photocatalyst for enhanced visible-light hydrogen evolution
Cheng R, et al.
Royal Society of Chemistry Advances, 6(23), 18990-18995 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service