Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800455

Sigma-Aldrich

Potassium bromide

AR, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

AR

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)

product line

Vetec

Assay

≥99.5%

form

crystalline powder

pH

5.0-6 (25 °C, 119 g/L)

mp

734 °C (lit.)

anion traces

chloride (Cl-): ≤0.3%

cation traces

Fe: ≤0.0003%
heavy metals (as Pb): ≤0.0002%

SMILES string

[K+].[Br-]

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

May be used for removal of peripheral membrane proteins.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zai-Chao Ma et al.
Marine biotechnology (New York, N.Y.), 17(4), 511-522 (2015-05-20)
In this study, in order to directly and efficiently convert inulin into pullulan, the INU1 gene from Kluyveromyces maximum KM was integrated into the genomic DNA and actively expressed in the high pullulan producer Aureobasidium melanogenum P16 isolated from the
Igor L Dalinger et al.
Chemistry, an Asian journal, 10(9), 1987-1996 (2015-06-23)
A new family of energetic compounds, nitropyrazoles bearing a trinitromethyl moiety at the nitrogen atom of the heterocycle, was designed. The desirable high-energy dense oxidizers 3,4-dinitro- and 3,5-dinitro-1-(trinitromethyl)pyrazoles were synthesized in good yields by destructive nitration of the corresponding 1-acetonylpyrazoles.
Sophie Rodrigues et al.
Probiotics and antimicrobial proteins, 7(1), 45-51 (2014-10-22)
Vibrio tapetis CECT4600 is a pathogenic Gram-negative bacterium causing the brown ring disease in the Manila clam Ruditapes philippinarum. This vibriosis is induced by bacterial attachment on the periostracal lamina, yielding a decalcification of the bivalve shell. As in many
Stephanie A DeJong et al.
Applied spectroscopy, 69(6), 733-748 (2015-05-09)
Derivatives are common preprocessing tools, typically implemented as Savitzky-Golay (SG) smoothing derivatives. This work discusses the implementation and optimization of fourth-order gap derivatives (GDs) as an alternative to SG derivatives for processing infrared spectra before multivariate calibration. Gap derivatives approximate
Thomas Trevethan et al.
Small (Weinheim an der Bergstrasse, Germany), 7(9), 1264-1270 (2011-04-13)
Modification and functionalization of the atomic-scale structure of insulating surfaces is fundamental to catalysis, self-assembly, and single-molecule technologies. Specially designed syn-5,10,15-tris(4-cyanophenylmethyl)truxene molecules can reshape features on an ionic KBr (001) surface. Atomic force microscopy images demonstrate that both KBr monolayer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service