Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800303

Sigma-Aldrich

Phenolphthalein solution

0.1 N

Synonym(s):

3,3-Bis(4-hydroxyphenyl)-1(3H)-isobenzofuranone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14O4
CAS Number:
Molecular Weight:
318.32
Beilstein:
284423
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

product line

Vetec

Quality Level

concentration

0.1 N

functional group

ester

SMILES string

Oc1ccc(cc1)C2(OC(=O)c3ccccc23)c4ccc(O)cc4

InChI

1S/C20H14O4/c21-15-9-5-13(6-10-15)20(14-7-11-16(22)12-8-14)18-4-2-1-3-17(18)19(23)24-20/h1-12,21-22H

InChI key

KJFMBFZCATUALV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Muta. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kwang-Seup Shin et al.
Food science and biotechnology, 28(4), 1027-1036 (2019-07-06)
The quality characteristics of commercial alcoholic beverages (yakju, diluted soju, and fruit wines) were evaluated by assessing pH, total acidity, acetaldehyde, SO2, and flavonoid content. pH level was the highest in diluted soju, and total acidity and flavonoid content were
Richa Pandey et al.
Biosensors & bioelectronics, 117, 493-500 (2018-07-10)
The ability to interact with plants, both to sense and to actuate, would open new opportunities for precision agriculture. These interactions can be achieved by using the plant as part of the sensing system. The present work demonstrates real-time monitoring
Ann M Dring et al.
Chemico-biological interactions, 188(3), 512-525 (2010-09-28)
Constitutive androstane receptor (CAR) and pregnane X receptor (PXR) are closely related orphan nuclear receptor proteins that share several ligands and target overlapping sets of genes involved in homeostasis and all phases of drug metabolism. CAR and PXR are involved
Pieter A Cohen et al.
Journal of general internal medicine, 27(1), 51-56 (2011-08-17)
Pai You Guo is a weight loss supplement manufactured in China and adulterated with the banned pharmaceutical products sibutramine and phenolphthalein. The US Food and Drug Administration (FDA) announced a voluntary recall of Pai You Guo in 2009, yet clinicians
Wasin Wongwilai et al.
Talanta, 81(3), 1137-1141 (2010-03-20)
A modification of a webcam camera for use as a small and low cost detector was demonstrated with a simple lab-on-chip reactor. Real time continuous monitoring of the reaction zone could be done. Acid-base neutralization with phenolphthalein indicator was used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service