Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800246

Sigma-Aldrich

Manganese(II) acetate tetrahydrate

LR, ≥99%

Synonym(s):

Manganous acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Mn · 4H2O
CAS Number:
Molecular Weight:
245.09
Beilstein:
3730728
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

LR

vapor pressure

<1 hPa ( 25 °C)

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

7 (20 °C, 50 g/L)

mp

>300 °C (lit.)

density

1.589 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.005%

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].CC(=O)O[Mn]OC(C)=O

InChI

1S/2C2H4O2.Mn.4H2O/c2*1-2(3)4;;;;;/h2*1H3,(H,3,4);;4*1H2/q;;+2;;;;/p-2

InChI key

CESXSDZNZGSWSP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renfa Liu et al.
Theranostics, 5(10), 1144-1153 (2015-08-19)
The integration of diagnostic and therapeutic functionalities into one nanoplatform shows great promise in cancer therapy. In this research, manganese (II) chelate functionalized copper sulfide nanoparticles were successfully prepared using a facile hydrothermal method. The obtained ultrasmall nanoparticles exhibit excellent
Kyu-Nam Jung et al.
Scientific reports, 5, 7665-7665 (2015-01-08)
Rechargeable metal-air batteries are considered a promising energy storage solution owing to their high theoretical energy density. The major obstacles to realising this technology include the slow kinetics of oxygen reduction and evolution on the cathode (air electrode) upon battery
Rusi et al.
PloS one, 10(7), e0129780-e0129780 (2015-07-15)
The composite metal oxide electrode films were fabricated using ex situ electrodeposition method with further heating treatment at 300°C. The obtained composite metal oxide film had a spherical structure with mass loading from 0.13 to 0.21 mg cm(-2). The structure
Qiangqiang Sun et al.
Journal of hazardous materials, 286, 276-284 (2015-01-16)
Comparative experiments were conducted to investigate the catalytic ability of MnO(x)/SBA-15 for the ozonation of clofibric acid (CA) and its reaction mechanism. Compared with ozonation alone, the degradation of CA was barely enhanced, while the removal of TOC was significantly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service