Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800241

Sigma-Aldrich

Magnesium chloride hexahydrate

LR, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

LR

product line

Vetec

Assay

≥98%

form

crystalline powder

pH

4.5-7

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Yang et al.
International journal of pharmaceutics, 471(1-2), 258-263 (2014-05-20)
Telmisartan (TEL) belongs to BCS class II (low solubility/high permeability) and exhibits the pH-dependent drug release. Since 3-aminopropyl functionalized magnesium phyllosilicate (aminoclay) can intercalate or adsorb the negatively charged molecules via the electrostatic interaction, TEL-aminoclay complex was synthesized to improve
Xingchu Gong et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 93, 260-266 (2015-04-25)
Brittleness is one of the important material properties that influences the success or failure of powder compaction. We have discovered that the reciprocal of diametrical elastic strain at fracture is the most suitable tablet brittleness indices (TBIs) for quantifying brittleness
Monica Mazzarino et al.
Analytica chimica acta, 837, 70-82 (2014-07-09)
This article describes a liquid chromatographic/tandem mass spectrometric method, based on the use of precursor ion scan as the acquisition mode, specifically developed to detect indole-derived cannabinoids (phenylacetylindoles, naphthoylindoles and benzoylindoles) in biological fluids (saliva, urine and blood). The method
Ikuo Yamamiya et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(9), 1485-1492 (2014-07-09)
Tegafur (FT), a prodrug of 5-fluorouracil, is a chiral molecule, a racemate of R- and S-isomers, and CYP2A6 plays an important role in the enantioselective metabolism of FT in human liver microsomes (R-FT > S-FT). This study examined the enantioselective
Heleen Kraan et al.
Vaccine, 33(17), 2030-2037 (2015-03-17)
In the near future oral polio vaccine (OPV) will be replaced by inactivated polio vaccine (IPV) as part of the eradication program of polio. For that reason, there is a need for substantial amount of safe and more affordable IPV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service