Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800235

Sigma-Aldrich

Lead(II) oxide

LR, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbO
CAS Number:
Molecular Weight:
223.20
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:

grade

LR

vapor pressure

10 mmHg ( 0 °C)

product line

Vetec

Assay

≥98%

form

powder

pH

9.9 (20 °C, 100 g/L)

mp

886 °C (lit.)

SMILES string

O=[PbH2]

InChI

1S/O.Pb

InChI key

YEXPOXQUZXUXJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Kidney,Blood

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Berashevich et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(7), 075803-075803 (2013-01-24)
The electronic properties of polycrystalline lead oxide consisting of a network of single-crystalline α-PbO platelets and the formation of native point defects in the α-PbO crystal lattice are studied using first-principles calculations. The results suggest that the polycrystalline nature of
Yin Wang et al.
Journal of colloid and interface science, 389(1), 236-243 (2012-10-16)
Lead(IV) oxide (PbO(2)) is a corrosion product on lead pipes used for drinking water distribution, and its dissolution can control lead release to drinking water. This study evaluated the adsorption of Pb(II) to PbO(2) and its impact on the dissolution
Ruqiang Zou et al.
Journal of the American Chemical Society, 132(51), 17996-17999 (2010-12-09)
A novel metal-organic replica of α-PbO(2) exhibits high capacity for capture of nerve agent surrogate.
Guoting Li et al.
Journal of environmental sciences (China), 23(6), 998-1003 (2011-11-10)
Beta-PbO2 electrodes were prepared by electro-deposition and characterized by scanning electron microscopy, X-ray diffraction, X-ray photoelectron spectroscopy, and linear sweep voltammetry. We confirmed pure (beta-PbO2 crystals were on the electrode and it had a high oxygen evolution potential. The photoactivity
Hui Lin et al.
Water research, 46(7), 2281-2289 (2012-03-03)
Electrochemical decomposition of environmentally persistent perfluorooctanoic acid (PFOA) in aqueous solution was investigated over Ti/SnO(2)-Sb, Ti/SnO(2)-Sb/PbO(2), and Ti/SnO(2)-Sb/MnO(2) anodes. The degradation of PFOA followed pseudo-first-order kinetics. The degradation ratios on Ti/SnO(2)-Sb, Ti/SnO(2)-Sb/PbO(2), and Ti/SnO(2)-Sb/MnO(2) anodes achieved 90.3%, 91.1%, and 31.7%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service