Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800089

Sigma-Aldrich

Bromophenol Blue

Powder

Synonym(s):

3′,3″,5′,5″-Tetrabromophenolsulfonphthalein, Bromphenol Blue Sultone Form

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H10Br4O5S
CAS Number:
Molecular Weight:
669.96
Beilstein:
61698
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Product Name

Bromophenol Blue,

product line

Vetec

form

powder

visual transition interval

3.0-4.6, yellow to purple

mp

273 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1c(Br)cc(cc1Br)C2(OS(=O)(=O)c3ccccc23)c4cc(Br)c(O)c(Br)c4

InChI

1S/C19H10Br4O5S/c20-12-5-9(6-13(21)17(12)24)19(10-7-14(22)18(25)15(23)8-10)11-3-1-2-4-16(11)29(26,27)28-19/h1-8,24-25H

InChI key

UDSAIICHUKSCKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ibrahim Alkatout et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 28(1), 69-79 (2014-06-21)
The prognosis of pancreatic neuroendocrine tumors is related to size, histology and proliferation rate. However, this stratification needs to be refined further. We conducted a proteome study on insulinomas, a well-defined pancreatic neuroendocrine tumor entity, in order to identify proteins
Julian A Zagalak et al.
RNA (New York, N.Y.), 21(12), 2132-2142 (2015-10-31)
Short interfering RNAs (siRNAs) are mediators of RNA interference (RNAi), a commonly used technique for selective down-regulation of target gene expression. Using an equimolar mixture of A, G, C, and U phosphoramidites during solid-phase synthesis, we introduced degenerate positions into
Dana Westphal et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(39), E4076-E4085 (2014-09-18)
The pivotal step on the mitochondrial pathway to apoptosis is permeabilization of the mitochondrial outer membrane (MOM) by oligomers of the B-cell lymphoma-2 (Bcl-2) family members Bak or Bax. However, how they disrupt MOM integrity is unknown. A longstanding model
Helen Karlberg et al.
The Journal of general virology, 96(Pt 3), 538-546 (2014-12-08)
Pathogenesis of viral haemorrhagic fevers is associated with alteration of vascular barrier function and haemorrhage. To date, the specific mechanism behind this is unknown. Programmed cell death and regulation of apoptosis in response to viral infection is an important factor
Srinita Banerjee et al.
The Journal of investigative dermatology, 135(1), 102-109 (2014-07-08)
House dust mites (HDMs) belong to the most potent indoor allergen sources worldwide and are associated with allergic manifestations in the respiratory tract and the skin. Here we studied the importance of the high-molecular-weight group 11 allergen from Dermatophagoides pteronyssinus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service