Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800038

Sigma-Aldrich

Ammonium carbonate

AR, ≥95%

Synonym(s):

Hartshorn salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2CO3
CAS Number:
Molecular Weight:
96.09
Beilstein:
4253487
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥95%
grade:
AR
form:
crystals

grade

AR

vapor density

2.7 (vs air)

product line

Vetec

Assay

≥95%

form

crystals

anion traces

sulfate (SO42-): ≤0.001%

cation traces

Fe: ≤0.0001%
heavy metals (as Pb): ≤0.0001%

SMILES string

N.N.OC(O)=O

InChI

1S/CH2O3.2H3N/c2-1(3)4;;/h(H2,2,3,4);2*1H3

InChI key

PRKQVKDSMLBJBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N N Mal'tseva et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 62(3), 56-65 (2000-08-10)
The intensification of activity of associative and symbiotic nitrogen fixation while using low amounts of ammonium-carbonate compounds, carbonic-ammonium salts, in particular, has been established. The main ways and mechanisms of the action of ammonium-carbonate salts as a new and perspective
Debabrata Nandi et al.
Journal of colloid and interface science, 454, 69-79 (2015-05-26)
Reduced-graphene-oxide based superparamagnetic nanocomposite (GC) was fabricated and applied for the remediation of Ni(II) from an aqueous medium. The as-prepared GC was extensively characterized by Raman, TEM, AFM, SEM-EDX, SQUID, and BET analyses. Quantitative immobilization of Ni(II) in an aqueous
Tomohisa Hasunuma et al.
Biotechnology for biofuels, 7(1), 493-493 (2015-02-05)
To ensure reliable sources of energy and raw materials, the utilization of sustainable biomass has considerable advantages over petroleum-based energy sources. Photosynthetic algae have attracted attention as a third-generation feedstock for biofuel production, because algae cultivation does not directly compete
Itsuo Murakami et al.
PloS one, 9(12), e115359-e115359 (2014-12-17)
Resveratrol (RESV) is a plant polyphenol, which is thought to have beneficial metabolic effects in laboratory animals as well as in humans. Following oral administration, RESV is immediately catabolized, resulting in low bioavailability. This study compared RESV metabolites and their
Mirosława Różycka et al.
PloS one, 9(12), e114308-e114308 (2014-12-10)
Fish otoliths, biominerals composed of calcium carbonate with a small amount of organic matrix, are involved in the functioning of the inner ear. Starmaker (Stm) from zebrafish (Danio rerio) was the first protein found to be capable of controlling the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service