Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800027

Sigma-Aldrich

Aluminum sulfate octadecahydrate

AR, ≥98%

Synonym(s):

Aluminum sulfate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2(SO4)3 · 18H2O
CAS Number:
Molecular Weight:
666.43
EC Number:
UNSPSC Code:
12352302

grade

AR

product line

Vetec

Assay

≥98%

form

solid

pH

2.5-4

anion traces

chloride (Cl-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%

SMILES string

O=S(O)(O)=O.[AlH2][AlH2].O

InChI

1S/2Al.3H2O4S.9H2O/c;;3*1-5(2,3)4;;;;;;;;;/h;;3*(H2,1,2,3,4);9*1H2/q2*+3;;;;;;;;;;;;/p-6

InChI key

KFCWRPFNNIEZRY-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service