Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V000976

Sigma-Aldrich

Azocarmine G

Synonym(s):

Acid Red 101, Rosinduline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H18N3NaO6S2
CAS Number:
Molecular Weight:
579.58
Colour Index Number:
50085
EC Number:
MDL number:
UNSPSC Code:
12352200

form

powder

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[O-]S(=O)(=O)c1ccc(Nc2cc3[n+](-c4ccccc4)c5ccccc5nc3c6ccc(cc26)S([O-])(=O)=O)cc1

InChI

1S/C28H19N3O6S2.Na/c32-38(33,34)20-12-10-18(11-13-20)29-25-17-27-28(22-15-14-21(16-23(22)25)39(35,36)37)30-24-8-4-5-9-26(24)31(27)19-6-2-1-3-7-19;/h1-17H,(H2,32,33,34,35,36,37);/q;+1/p-1

InChI key

LUERODMRBLNCFK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Duval et al.
PLoS neglected tropical diseases, 9(2), e0003489-e0003489 (2015-02-27)
Schistosomiasis is the second-most widespread tropical parasitic disease after malaria. Various research strategies and treatment programs for achieving the objective of eradicating schistosomiasis within a decade have been recommended and supported by the World Health Organization. One of these approaches

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service