Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52722-U

Supelco

Discovery® DSC-8 SPE Tube

bed wt. 10 g, volume 60 mL, pk of 16

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

material

polypropylene tube

composition

bed wt., 10 g

packaging

pk of 16

extent of labeling

9% C loading

technique(s)

solid phase extraction (SPE): suitable

surface area

480 m2/g

volume

60 mL

matrix

Silica gel base material (irregularly shaped, acid washed)

matrix active group

C8 (octyl) bonding, monomerically bonded

particle size

50 μm

pore size

0.9 mL/g pore volume
70 Å pore diameter

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Reversed-phase
Sample Matrix Compatibility: Aqueous solutions (biological fluids, water)

  • Monomerically bonded, octyl (9 %C), endcapped; lower carbon content than DSC-18Lt
  • Used to elute very large hydrophobic molecules too strongly retained on DSC-18 or DSC-18Lt
  • Use this less retentive phase for the rapid release of hydrophobic molecules using weaker organic solvents at lower volumes
  • Inorganic buffers of sufficient ionic strength may be used for elution

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service