Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

509787

SGE Microvolume Syringe for Rheodyne® and Valco® Valves, Fixed Needle

10F-LC, volume 10 μL, needle size 22 ga, needle L × O.D. 2 in. × 0.7112 mm (0.028 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

blunt tip stainless steel needle (point style 3)

needle size

22 ga

feature

Fixed needle

packaging

pkg of 1 ea

manufacturer/tradename

SGE 002301

needle L × O.D.

2 in. × 0.7112 mm (0.028 in.)

volume

10 μL

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Rheodyne is a registered trademark of IDEX Health & Science, LLC
Valco is a registered trademark of Valco Instruments Co. Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service