Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47660-U

Supelco

Dibromoacetic acid solution

certified reference material, 1000 μg/mL in methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 552.2

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in methyl tert-butyl ether

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C2H2Br2O2/c3-1(4)2(5)6/h1H,(H,5,6)

InChI key

SIEILFNCEFEENQ-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingxue Sun et al.
Se pu = Chinese journal of chromatography, 24(3), 298-301 (2006-08-26)
The combined solid phase extraction (SPE)-ion chromatography (IC) method was developed for the analysis of trace haloacetic acids (HAAs) in drinking water. The tested HAAs included monochloroacetic acid (MCAA), dichloroacetic acid (DCAA), trichloroacetic acid (TCAA), monobromoacetic acid (MBAA) and dibromoacetic
National Toxicology Program technical report series, (537)(537), 1-320 (2007-06-08)
Dibromoacetic acid is a water disinfection by-product. Dibromoacetic acid was nominated to the National Toxicology Program by the United States Environmental Protection Agency for toxicity and carcinogenicity studies in rats and mice because of widespread human exposure and because a
Jerome M Goldman et al.
Reproductive toxicology (Elmsford, N.Y.), 23(4), 541-549 (2007-04-10)
The disinfection by-product dibromoacetic acid (DBA) has been found in female rats to increase circulating concentrations of both estradiol (E2) and estrone (E1). This effect is apparently due, at least in part, to a suppression in hepatic catabolism. The present
Ashley S Murr et al.
Reproductive toxicology (Elmsford, N.Y.), 20(1), 73-80 (2005-04-06)
Elevated gavage exposures to the drinking water disinfection by-product dibromoacetic acid (DBA) have been found to disrupt estrous cyclicity in the rat and induce increases in estradiol concentrations in both cycling (day of estrus) and ovariectomized/estradiol-implanted females. The present study
Eric R V Dickenson et al.
Environmental science & technology, 42(9), 3226-3233 (2008-06-05)
While it is known that resorcinol- and phenol-type aromatic structures within natural organic matter (NOM) react during drinking water chlorination to form trihalomethanes (THMs), limited studies have examined aliphatic-type structures as THM and haloacetic acid (HAA) precursors. A suite of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service