Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11034AST

Supelco

Astec® CHIROBIOTIC® V Chiral (5 μm) HPLC Columns

L × I.D. 25 cm × 10 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52
L × I.D.:
25 cm × 10 mm
particle size:
5 μm
matrix active group:
vancomycin phase
pore size:
100 Å
matrix:
High-purity silica gel particle platform
fully porous particle

Product Name

Astec® CHIROBIOTIC® V Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm

material

stainless steel column

Agency

suitable for USP L88

description

HPLC Column

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-45 °C temperature
241 bar pressure (3500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

25 cm × 10 mm

matrix

High-purity silica gel particle platform
fully porous particle

matrix active group

vancomycin phase

particle size

5 μm

pore size

100 Å

operating pH

3.5-7.0

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

Neutral molecules, amides, acids, esters and amines show considerable enantioselectivity on these vancomycin-based CSPs. A wide variety of secondary and tertiary amines have been separated on the CHIROBIOTIC® V in the polar ionic mode. The CHIROBIOTIC® V has demonstrated many of the separation characteristics of protein-based stationary phases, but with exceptional stability and much higher sample capacity. Some chiral analytes have been resolved that have not been reported separated on any other chiral stationary phase. CHIROBIOTIC® V and V2 differ in their bonding chemistry the pore size of the support particle, giving them different selectivity and preparative capacity.

  • Bonded phase: Vancomycin
  • Operating pH range: 3.5 - 7.0
  • Particle diameter: 5, 10 or 16 μm
  • Pore size: 100 Å (CHIROBIOTIC® V) or 200 Å (CHIROBIOTIC® V2)

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Bagnall et al.
Journal of chromatography. A, 1249, 115-129 (2012-07-04)
This paper presents and compares for the first time two chiral LC-QTOF-MS methodologies (utilising CBH and Chirobiotic V columns with cellobiohydrolase and vancomycin as chiral selectors) for the quantification of amphetamine, methamphetamine, MDA (methylenedioxyamphetamine), MDMA (methylenedioxymethamphetamine), propranolol, atenolol, metoprolol, fluoxetine
Analysis of the enantiomers of citalopram and its demethylated metabolites using chiral liquid chromatography
Kosel, M., et al.
Journal of Chromatography. B, Biomedical Applications, 719 (1-2), 234-238 (1998)
HPLC separation technique for analysis of bufuralol enantiomers in plasma and pharmaceutical formulations using a vancomycin chiral stationary phase and UV detection
Hefnawy, Mohamed M., et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 856 (1-2), 328-336 (2007)
Ana R Ribeiro et al.
Chemosphere, 95, 589-596 (2013-11-05)
Microbial degradation is the most important process to remove organic pollutants in Waste Water Treatment Plants. Regarding chiral compounds this process is normally enantioselective and needs the suitable analytical methodology to follow the removal of both enantiomers in an accurate
Hisham Hashem et al.
Journal of chromatography. A, 1218(38), 6727-6731 (2011-08-23)
In this study, a method for enantioseparation of terbutaline and salbutamol was established using Chirobiotic V column as a stationary phase. Polar ionic mode applying mobile phase containing ammonium nitrate in 100% ethanol, pH 5.1 was found to give the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service