Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

ZRB3127

Sigma-Aldrich

Anti-CHD4 Antibody, clone 1C17 ZooMAb® Rabbit Monoclonal

newenhanced validation

recombinant, expressed in HEK 293 cells

Synonym(s):

ATP-dependent helicase CHD4, CHD-4, Chromodomain-helicase-DNA-binding protein 4, Mi-2 autoantigen 218 kDa protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C17, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 218 kDa
observed mol wt ~240 kDa

purified by

using Protein A

species reactivity

mouse, human, rat

species reactivity (predicted by homology)

bovine, sheep, monkey, feline, horse

packaging

antibody small pack of 25

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... CHD4(1108)

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C17 is a ZooMAb® rabbit recombinant monoclonal antibody that detects CHD4. It targets an epitope within 21 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the N-terminal half of human CHD4.

Application

Quality Control Testing

Evaluated by Western Blotting in Raji cell nuclear extract.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected CHD4 in Raji cell nuclear extract.

Tested Applications

Western Blotting Analysis: A 1:1,000-10,000 dilution from a representative lot detected CHD4 in lysates from C6 and NIH/3T3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected CHD4 in human stomach cancer tissue sections and human small intestine tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CHD4 in NIH/3T3 cells.

Affinity Binding Assay: A representative lot of this antibody bound with CHD4 peptide at least ten thousand-fold (10,000X) higher than with the non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Chromodomain-helicase-DNA-binding protein 4 (UniProt: Q14839; EC:3.6.4.12; also known as CHD-4, ATP-dependent helicase CHD4, Mi-2 autoantigen 218 kDa protein, Mi2-beta) is encoded by the CHD4 gene (Gene ID: 1108) in human. CHD4 is a member of the CHD family of proteins, which are characterized by the presence of chromo domains, helicase domains, and DNA-binding domains. CHD4 is a central component of the nucleosome remodeling and deacetylase (NuRD) complex, regulating gene expression, involved in early B cell development, in neuronal differentiation and the repair of DNA double-strand breaks by facilitating chromatin relaxation at damage sites. CHD4 is expressed in various tissues, with particularly high during embryonic development. CHD4 has been implicated in several diseases. Mutations and dysregulation of CHD4 are associated with various cancers, including breast, ovarian, and colorectal cancers. It is linked to intellectual disabilities and developmental delay, highlighting its importance in brain development. Aberrant CHD4 activity can lead to immune dysregulation and contribute to autoimmune diseases. CHD4 dysfunction has been associated with congenital heart defects and other cardiovascular anomalies. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Hagman, J. R., (2022) Immunological Reviews, 305. 29 42)

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service