Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

ZRB2665

Sigma-Aldrich

Anti-TGF-β1 Antibody, clone 4E3 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

TGFB, Transforming growth factor beta-1, Transforming growth factor beta-1 proprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4E3, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 44.33 kDa
observed mol wt ~13 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

horse, bovine, monkey, porcine, rat, nonhuman primates, goat, sheep

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... TGFB1(7040)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 4E3 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects TGF-beta-1. It targets an epitope within 18 amino acids from the C-terminal region of the protein.

Immunogen

KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of human TGF- β1.

Application

Quality Control Testing

Evaluated by Western Blotting in TGF-β1 recombinant protein.

Western Blotting Analysis: A 1:50,000 dilution from a representative lot detected TGF-β1 in recombinant fragment from human TGF-β1 protein .

Tested Applications

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected TGF-β1 in HEK-293 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected TGF-β1 in human bone marrow and human stomach tissue sections.

Flow Cytometry Analysis (FC): 1 µg from a representative lot detected TGF-β1 in one million HEK-293 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Transforming growth factor beta-1 proprotein (UniProt: P01137) is encoded by the TGFB1 (also known as TGFB) gene (Gene ID: 7040) in human. Transforming growth factor beta-1 proprotein is synthesized with a signal peptide (aa 1-29) and is cleaved in the Golgi apparatus by FURIN to form two chains; Latency-associated peptide (LAP) (aa 30-278) and Transforming growth factor beta-1 (TGF-beta-1) (aa 279-390), which remain non-covalently linked, rendering TGF-beta-1 inactive. These 2 chains, LAP and TGF-beta-1 constitutes the regulatory and active subunit of TGF-beta-1, respectively. TGFB1 belongs to the TGF-β superfamily of secreted proteins (primarily localized in the extracellular matrix once secreted) and is involved in a variety of signaling pathways that involve serine/threonine protein kinases. It is widely expressed but tightly regulated at both the transcriptional and post-transcriptional levels. It is produced by many cell types, including macrophages, endothelial cells, fibroblasts, and epithelial cells and is involved in cell and tissue differentiation, proliferation, inflammation, migration of monocytes, neutrophils and chemotaxis. TGF-β acts through binding to three transmembrane receptors in mammals, type I, type II and type III receptors. The important pathways regulated by TGF-β are the SMAD, BMP, MAPK and PI3K/Akt pathways. TGFB1 is among the most potent fibrogenic cytokines inducing organ fibrosis by promoting the formation of myofibroblasts. TGFB1 is a key player in the development of fibrotic diseases such as pulmonary fibrosis, liver cirrhosis, and kidney fibrosis, where it stimulates the deposition of extracellular matrix components, leading to tissue scarring. Deregulation of TGF-β signaling has been implicated in many inflammation and autoimmune diseases.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service