Skip to Content
MilliporeSigma
All Photos(12)

Key Documents

ZRB1695

Sigma-Aldrich

Anti-NEDD4 Antibody, clone 1D20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D20, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 149.11 kDa
observed mol wt ~160 kDa

purified by

using Protein A

species reactivity

human, rat

species reactivity (predicted by homology)

bovine, horse, canine, monkey, porcine, feline

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NEDD4(4734)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1D20 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects NEDD4. It targets an epitope within 16 amino acids from the internal region.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the internal region of human NEDD4.

Application

Quality Control Testing

Evaluated by Western Blotting in Rat Kidney tissue lysate.

Western Blotting Analysis: A 1:3,000 dilution of this antibody detected NEDD4 in Rat Kidney tissue lysate.

Tested applications

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected NEDD4 in human cerebral cortex and human kidney tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound NEDD4 with a KD of 5.3 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

E3 ubiquitin-protein ligase NEDD4 (UniProt: P46934; also known as EC:2.3.2.26, Cell proliferation-inducing gene 53 protein, HECT-type E3 ubiquitin transferase NEDD4, Neural precursor cell expressed developmentally down-regulated protein 4, NEDD-4) is encoded by the NEDD4 (also known as KIAA0093, NEDD4-1, PIG53) gene (Gene ID: 4734) in human. NEDD4 is a peripheral membrane protein that is recruited to the plasma membrane by GRB10. Once complexed with GRB10 and IGF1R, it follows IGF1R internalization and remains associated with early endosomes. NEDD4 can accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates. It is shown to specifically ubiquitinate Lysine 63 in target proteins. It also monoubiquitinates IGF1R at multiple sites, thus leading to receptor internalization and degradation in lysosomes. NEDD4 is reported to polyubiquitinate the epithelial sodium channel and any mutations that disrupt association between NEDD4 and sodium channel can lead to Liddle s syndrome, a hyperaldosteronism-like hypertension disease. NEDD4 can interact with viral proteins that contain a late-budding motif P-P-P-Y and this interaction is believed to be essential for viral particle budding of many retroviruses, like HTLV-1 Gag, and MLV Gag. NEDD4 contains four WW domains (WW 1-4; aa 610-643; 767-800; 840-873; and 892-925) and one HECT domain (aa 984-1318). The WW domains mediate its interaction with PPxY motif-containing proteins. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Maspero, E., et al. (2013). Nat. Struct. Mol. Biol. 20(6); 696-701; Rauch, S., and Martin-Serrano, J. (2011). J. Virol. 85(7); 3546-3556).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service