Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

WH0375307M1

Sigma-Aldrich

Monoclonal Anti-MGC50811 antibody produced in mouse

clone 2C3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-hypothetical gene supported by BC052750

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
clone:
2C3, monoclonal
species reactivity:
human
citations:
technique(s):
indirect ELISA: suitable

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG1κ

GenBank accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Related Categories

Immunogen

MGC50811 (AAH52750, 1 a.a. ~ 387 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSSKVYSTGSRAKDHQPSGPECLPLPEANAEAIDFLSSLHKEELQMLFFSETLAMVSDTGEPQGELTIEVQRGKYQEKLGMLTYCLFVHASSRGFLDKMLCGNSLLGYLSEKLELMEQHSQDFIKFLILPMERKMSLLKQDDQLAVTRSIKEGEEVKTGVTSFPWSSIKGFISEAANLVLLRVMAWRRMVPSNARFLTLDTEGKLCYLTYQNLGFQTIQVDHQQAEVFIVEQTVHAEEGIPMSCQYYLLSDGHLAKRIQVGSPGCCIITKMPILREEDEIEPRPVFEKKPLVWEEDMELYSKFLDRKEELRLGHASYLRQHPEAHALISDFLLFLLLRQPEDVVTFAAEFFGPFDPWRPSSPALGSSHRPNPFRSLEPEGDARSGAA

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service