Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

WH0246778M1

Sigma-Aldrich

Monoclonal Anti-IL27 antibody produced in mouse

clone 3F12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-IL27, Anti-IL27p28, Anti-IL30, Anti-MGC71873, Anti-interleukin 27, Anti-p28

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
clone:
3F12, monoclonal
application:
ELISA (i)
IP
WB
species reactivity:
human
technique(s):
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL
citations:
2

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL27(246778)

General description

The protein encoded by this gene is one of the subunits of a heterodimeric cytokine complex. This protein is related to interleukin 12A (IL12A). It interacts with Epstein-Barr virus induced gene 3 (EBI3), a protein similar to interleukin 12B (IL12B), and forms a complex that has been shown to drive rapid expansion of naive but not memory CD4(+) T cells. The complex is also found to synergize strongly with interleukin 12 to trigger interferon gamma (IFNG) production of naive CD4(+) T cells. The biological effects of this cytokine are mediated by the class I cytokine receptor (WSX1/TCRR). (provided by RefSeq)

Immunogen

IL27 (NP_663634, 177 a.a. ~ 243 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
RKGLLPGALGSALQGPAQVSWPQLLSTYRLLHSLELVLSRAVRELLLLSKAGHSVWPLGFPTLSPQP

Biochem/physiol Actions

Interleukin-27 (IL27) has a role in immune responses. It aids in the secretion of interferon-γ and the activation of natural killer cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng Zhang et al.
Medical science monitor : international medical journal of experimental and clinical research, 21, 2505-2513 (2015-08-26)
Many epidemiology studies have indicated that several functional polymorphisms of the IL-27 gene may contribute to individual susceptibility to cancer. Nevertheless, the data arising from these studies were inconclusive. Therefore, we conducted the current meta-analysis aiming to elucidate the effects
Zhengting Wang et al.
International journal of clinical and experimental pathology, 7(12), 8952-8957 (2015-02-13)
To investigate the association of three polymorphisms in the Interleukin-27 (IL-27) gene with CD risk in Chinese population. This case-control study involved 312 CD patients and 479 age- and sex-matched healthy controls. Genotyping was performed using PCR-LDR method. Data were
Ha-Neul Jeong et al.
Journal of neuroimmunology, 288, 120-122 (2015-11-05)
Myasthenia gravis (MG)(1) is an autoimmune disease directed at the neuromuscular junction, and cytokines are thought to contribute to its immunopathogenesis. Interleukin-27 (IL-27)(2) plays a complex and pleiotropic role in immune responses associated with T helper cells. To assess the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service