Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

WH0023054M1

Sigma-Aldrich

Monoclonal Anti-NCOA6 antibody produced in mouse

clone 2D7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-AIB3, Anti-ASC2, Anti-KIAA0181, Anti-NRC, Anti-PRIP, Anti-RAP250, Anti-TRBP, Anti-nuclear receptor coactivator 6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
clone:
2D7, monoclonal
application:
ELISA (i)
IF
WB
species reactivity:
human
technique(s):
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL
citations:
2

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2D7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NCOA6(23054)

General description

The protein encoded by this gene is a transcriptional coactivator that can interact with nuclear hormone receptors to enhance their transcriptional activator functions. The encoded protein has been shown to be involved in the hormone-dependent coactivation of several receptors, including prostanoid, retinoid, vitamin D3, thyroid hormone, and steroid receptors. The encoded protein may also act as a general coactivator since it has been shown to interact with some basal transcription factors, histone acetyltransferases, and methyltransferases. (provided by RefSeq)

Immunogen

NCOA6 (NP_054790, 1954 a.a. ~ 2063 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VGSHPELLPSIAPSQNLVSKETSTTALQASVARPELEVNAAIVSGQSSEPKEIVEKSKIPGRRNSRTEEPTVASESVENGHRKRSSRPASASSSTKDITSAVQSKRRKSK

Biochem/physiol Actions

NCOA6 (Nuclear receptor coactivator 6) interacts with nuclear receptors in ligand-dependent or ligand-enhanced dependent manner to regulate nuclear receptor-mediated mammalian gene expression. The activation domain helps to increase the transcriptional activity of several nuclear receptors. It forms a dimer by binding to the liganded receptors, which lead to a structural change in NRC activation. It has also been reported that NCOA6 highly influences the binding of CREB-binding protein (CBP) during transcriptional activity of nuclear hormone receptors.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Mahajan et al.
Molecular and cellular biology, 20(14), 5048-5063 (2000-06-24)
We describe the cloning and characterization of a new family of nuclear receptor coregulators (NRCs) which modulate the function of nuclear hormone receptors in a ligand-dependent manner. NRCs are expressed as alternatively spliced isoforms which may exhibit different intrinsic activities
F Caira et al.
The Journal of biological chemistry, 275(8), 5308-5317 (2000-02-22)
Ligand-induced transcriptional activation of gene expression by nuclear receptors is dependent on recruitment of coactivators as intermediary factors. The present work describes the cloning and characterization of RAP250, a novel human nuclear receptor coactivator. The results of in vitro and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service