Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V5626

Sigma-Aldrich

Val-Tyr

>98% (TLC), suitable for ligand binding assays

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2O4
CAS Number:
Molecular Weight:
280.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

product name

Val-Tyr,

Assay

>98% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

OC([C@@H](NC([C@@H](N)C(C)C)=O)CC1=CC=C(O)C=C1)=O

InChI

1S/C14H20N2O4/c1-8(2)12(15)13(18)16-11(14(19)20)7-9-3-5-10(17)6-4-9/h3-6,8,11-12,17H,7,15H2,1-2H3,(H,16,18)(H,19,20)

InChI key

VEYJKJORLPYVLO-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jocksan I Morales-Camacho et al.
Applied microbiology and biotechnology (2018-09-14)
The insertion of peptides is a biotechnology tool widely used to improve the nutraceutical properties of proteins. Because the effect of these insertions in protein stability and function is difficult to predict, it should be determined experimentally. In this study
Mitsuru Tanaka et al.
Bioscience, biotechnology, and biochemistry, 70(9), 2292-2295 (2006-09-09)
In this study, we found that antihypertensive di-peptide Val-Tyr (VY) showed a vascular relaxation effect in KCl-induced contraction of thoracic aorta rings from 18-week-old spontaneously hypertensive rats among di-peptides of VY, Ile-Tyr, and Tyr-Val irrespective of their angiotensin I-converting enzyme
Takao Ueno et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 21(8), 997-1000 (2005-08-27)
A double column-switching HPLC method with naphthalene-2,3-dialdehyde (NDA) was applied for determination of two plasma antihypertensive peptides, Val-Tyr (VY) and Ile-Val-Tyr (IVY). After a first separation on a Phe-ODS column, double heart-cuts of the retention time corresponding to NDA-VY and
Mitsuru Tanaka et al.
Scientific reports, 9(1), 5769-5769 (2019-04-10)
Apart from nutrients required for the brain, there has been no report that naturally occurring peptides can cross the blood-brain barrier (BBB). The aim of this study was to identify the BBB-transportable peptides using in situ mouse perfusion experiments. Based
Lieselot Vercruysse et al.
Peptides, 29(2), 261-267 (2008-01-29)
Antihypertensive peptides received much interest over the last decade. These peptides are known to be angiotensin converting enzyme (ACE) inhibitors in vitro, but the actual antihypertensive mechanisms in vivo are still unclear. In this research, we used rat aortic rings

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service