Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

U6385

Sigma-Aldrich

Ubiquitin-Specific Protease 14 human

recombinant, expressed in Sf9 cells, ≥75% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in Sf9 cells

Quality Level

Assay

≥75% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

57 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

General description

Human USP14 (Ubiquitin-Specific Protease 14) or TGT (tRNA-guanine transglycosylase), amino acids 2-494 (end) with an N-terminal FLAG-tag, expressed in a baculovirus-infected Sf9 cell expression system.
Ubiquitin-Specific Protease 14 is a deubiquitinating protein responsible for the removal of ubquitin tags attached to proteins by α•NH-peptide bonds and the ε-NH-isopeptide bond.
USP14 has been shown to be phosphorylated by AKT which is required for protein stability and nuclear localization.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Physical form

Solution in 40 mM Tris-HCl, 110 mM NaCl, 2.2 mM KCl, 80 ng/μl Flag peptide, 3 mM DTT, and 20% Glycerol at pH 8.0.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long Zhang et al.
Nature cell biology, 14(7), 717-726 (2012-06-19)
The stability and membrane localization of the transforming growth factor-β (TGF-β) type I receptor (TβRI) determines the levels of TGF-β signalling. TβRI is targeted for ubiquitylation-mediated degradation by the SMAD7-SMURF2 complex. Here we performed a genome-wide gain-of-function screen and identified
Atsushi Nagai et al.
Biochemical and biophysical research communications, 379(4), 995-1000 (2009-01-13)
Accumulation of unfolded proteins within the endoplasmic reticulum (ER) lumen induces ER stress. Eukaryotic cells possess the ER quality control systems, the unfolded protein response (UPR), to adapt to ER stress. IRE1alpha is one of the ER stress receptors and
Yi-Chen Chen et al.
RNA (New York, N.Y.), 16(5), 958-968 (2010-04-01)
The eukaryotic tRNA-guanine transglycosylase (TGT) has been reported to exist as a heterodimer, in contrast to the homodimeric eubacterial TGT. While ubiquitin-specific protease 14 (USP14) has been proposed to act as a regulatory subunit of the eukaryotic TGT, the mouse
Ping-Chung Chen et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(35), 10909-10919 (2009-09-04)
Dysfunction of the ubiquitin proteasome system (UPS) has been implicated in the pathogenesis of many neurological diseases, including Alzheimer's, spinocerebellar ataxia, and several motor neuron diseases. Recent research indicates that changes in synaptic transmission may play a critical role in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service