Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T9001

Sigma-Aldrich

Tricaprylin

Sigma Grade, ~99%

Synonym(s):

1,2,3-Trioctanoylglycerol, Glyceryl tricaprylate

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352202

grade

Sigma Grade

Quality Level

Assay

~99%

density

0.95 g/mL at 25 °C (lit.)

storage temp.

−20°C

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

392.4 °F

Flash Point(C)

200.2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service