Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T8825

Sigma-Aldrich

Anti-phospho-Tryptophan Hydroxylase (pSer58) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution, sufficient for 10 blots

Synonym(s):

Anti-TPH

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

usage

sufficient for 10 blots

species reactivity

Xenopus, rat, human, mouse

technique(s)

dot blot: 1:1,000
western blot: 1:1000 using human dorsal Raphe nucleus

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer58)

Gene Information

human ... TPH1(7166)
mouse ... Tph1(21990)
rat ... Tph1(24848)

Immunogen

synthetic phosphopeptide corresponding to amino acid residues surrounding phosphorylated serine 58 of tryptophan hydroxylase.

Target description

Tryptophan hydroxylase (TPH) catalyzes the 5-hydroxylation of tryptophan, which is the first step in the biosynthesis of indoleamines (serotonin and melatonin)

Physical form

Solution in 100 μL in 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 μg per mL BSA, and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service