Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6172

Sigma-Aldrich

IL-4 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

B-cell stimulatory factor 1 (BSF-1), BCGF, Binetrakin, Lymphocyte stimulatory factor 1, Pitrakinra

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

(14 and 19 kDa, monomer)

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IL4(3565)

General description

Interleukin-4 (IL-4) is a pleiotropic cytokine and a potent lymphoid cell growth factor. It is produced by activated T cells and is a ligand for interleukin 4 receptor. The gene encoding this protein is localized on human chromosome 5q31.1. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.

Biochem/physiol Actions

Interleukin-4 (IL-4) stimulates the growth and survivability of certain B cells and T cells. The interleukin 4 receptor also binds to interleukin-13 (IL-13), which may contribute to many overlapping functions of this cytokine and IL-13. STAT6 (signal transducer and activator of transcription-6) has been shown to play a central role in mediating the immune regulatory signal of this cytokine. This protein exhibits anti-inflammatory responses and has a role in immune processes by protecting against intracellular pathogens.

Physical form

Lyophilized from a solution of 20 mM Tris-HCl, pH 7.4, with 300 mM NaCl.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Reconstitute in sterile PBS containing 0.1% endotoxin-free recombinant human serum albumin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interleukin 4 mediates autocrine growth of helper T cells after antigenic stimulation.
Fernandez-Botran R
Proceedings of the National Academy of Sciences of the USA, 83(24), 9689-9693 (1986)
Interleukin 4 inhibition as a potential therapeutic in pemphigus.
Tavakolpour S and Tavakolpour V
Cytokine, 77, 189-195 (2016)
D G Marsh et al.
Science (New York, N.Y.), 264(5162), 1152-1156 (1994-05-20)
Sib-pair analysis of 170 individuals from 11 Amish families revealed evidence for linkage of five markers in chromosome 5q31.1 with a gene controlling total serum immunoglobulin E (IgE) concentration. No linkage was found between these markers and specific IgE antibody
Masanori Kobayashi et al.
Cancer immunology, immunotherapy : CII, 63(8), 797-806 (2014-04-30)
Dendritic cell (DC)-based cancer vaccines may have a significant benefit to patients with advanced pancreatic cancer. However, variations among clinical studies make it difficult to compare clinical outcomes. Here, we identified factors that determined the clinical benefits by analyzing data
Michelle A Favila et al.
Journal of immunology (Baltimore, Md. : 1950), 192(12), 5863-5872 (2014-05-09)
Leishmania major-infected human dendritic cells (DCs) exhibit a marked induction of IL-12, ultimately promoting a robust Th1-mediated response associated with parasite killing and protective immunity. The host cell transcription machinery associated with the specific IL-12 induction observed during L. major

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service