Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5170

Sigma-Aldrich

CaMKPase, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FEM-2, KIAA0015, POPX2, PPM1F, hFEM-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~74 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PPM1F(9647)

General description

CaMKPase is a member of the PP2C family of Ser/Thr protein phosphatases that dephosphorylate and regulate the multifunctional Ca2+/calmodulin-dependent protein kinases (CaMKs). The multifunctional CaMKs mediate cellular responses induced by increases in second messenger Ca2+ and have been implicated in the control of synaptic transmission, gene transcription, cell growth and contraction of cardiac and smooth muscles. Overexpression of CaMKPase has been shown to mediate caspase-dependent apoptosis. CaMKPase can also interact with the Rho guanine nucleotide exchange factors (PIX) thereby blocking the effects of p21-activated kinase 1 (PAK1).

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atsuhiko Ishida et al.
Archives of biochemistry and biophysics, 408(2), 229-238 (2002-12-05)
Ca(2+)/calmodulin-dependent protein kinase phosphatase (CaMKPase) dephosphorylates and regulates multifunctional Ca(2+)/calmodulin-dependent protein kinases (CaMKs). One of the prominent features of CaMKPase is stimulation of phosphatase activity by polycations such as poly-L-lysine (poly(Lys)). Using various polycations, basicity and molecular weight of the
I Kameshita et al.
FEBS letters, 456(2), 249-252 (1999-08-24)
Ca2+/calmodulin-dependent protein kinase phosphatase (CaMKPase) is a protein phosphatase which dephosphorylates autophosphorylated Ca2+/calmodulin-dependent protein kinase II (CaMKII) and deactivates the enzyme (Ishida, A., Kameshita, I. and Fujisawa, H. (1998) J. Biol. Chem. 273, 1904-1910). In this study, a phosphorylation-dephosphorylation relationship

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service