Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5111

Sigma-Aldrich

p33ING1, His tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ING1, p24ING1c, p33, p33ING1b, p47, p47ING1a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~38 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ING1(3621)

General description

p33ING1 displays characteristics of a tumour suppressor protein and can induce cell growth arrest and apoptosis. p33ING1 is a nuclear protein and acute expression of p33 ING1 inhibits cell growth. p33ING1 physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Mutation of p33ING1 gene occurs in neuroblastoma cells and reduced expression is observed in some breast cancer cell lines. It has been proposed that p33ING1 can act as a potent growth regulator in normal and in established cells and plays a role as a candidate tumour suppressor gene whose inactivation may contribute to the development of cancers.

Physical form

Supplied in 50mM MOPS, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suppression of the novel growth inhibitor p33ING1 promotes neoplastic transformation
Garkavtsev
Nature genetics, 23(3), 373-373 (1999-11-05)
Frauke Goeman et al.
Molecular and cellular biology, 25(1), 422-431 (2004-12-17)
ING1 was identified as an inhibitor of growth and has been described as a tumor suppressor. Furthermore, the expression of ING1 is induced in senescent cells and antisense ING1 extends the proliferative life span of primary human fibroblasts. Cooperation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service