Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2044

Sigma-Aldrich

PPAR, β human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

FAAR, MGC3931, NR1C2, NUC1, NUCI, NUCII, PPARB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~52 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

350 μg/mL

technique(s)

electrophoretic mobility shift assay: suitable

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PPARD(5467)

Biochem/physiol Actions

There is evidence that a group of closely related nuclear receptors, called peroxisome proliferator-activated receptors (PPARs), may be involved in chronic diseases such as diabetes, obesity, artherosclerosis and cancer. The PPARs were first cloned as the nuclear receptors that mediate the effects of synthetic compounds called peroxisome proliferators on gene transcription. It soon became clear that eicosanoids and fatty acids can also regulate gene transcription through PPARs. They bind a specific element in the promoter region of target genes only as a heterodimer with the receptor for 9-cis retinoic acid, RXR (retinoid X receptor). Binding of the ligand of either receptor can activate the complex, but binding of both ligands simultaneously is more potent. Three PPAR isotypes have been identified: α, β (also called NUC1) and γ. PPAR α is expressed most in brown adipose tissue and liver, then kidney, heart and skeletal muscle. PPAR β is found in many tissues but the highest expression is in the gut, kidney and heart. PPAR γ is mainly expressed in adipose tissue, and to a lesser extent in colon, the immune system and the retina. The target genes of PPAR α are a relatively homogenous group of genes that participate in aspects of lipid catabolism such as fatty acid uptake through membranes, fatty acid binding in cells, fatty acid oxidation (in microsomes, peroxisomes and mitochondria) and lipoprotein assembly and transport.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T C He et al.
Cell, 99(3), 335-345 (1999-11-11)
PPARB was identified as a target of APC through the analysis of global gene expression profiles in human colorectal cancer (CRC) cells. PPARdelta expression was elevated in CRCs and repressed by APC in CRC cells. This repression was mediated by
Peroxisome proliferator-activated receptors: nuclear control of metabolism.
B Desvergne et al.
Endocrine reviews, 20(5), 649-688 (1999-10-26)
S Kersten et al.
Nature, 405(6785), 421-424 (2000-06-06)
In developed societies, chronic diseases such as diabetes, obesity, atherosclerosis and cancer are responsible for most deaths. These ailments have complex causes involving genetic, environmental and nutritional factors. There is evidence that a group of closely related nuclear receptors, called

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service