Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0729

Sigma-Aldrich

PCTK3 (CDK18)/CYCLINY, Active

PRECISIO® Kinase, recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

35-55 nmol/min·mg (radiometric)

mol wt

~64 kDa (Cyclin Y)
~76 kDa (PCTK3)

concentration

0.1 μg/μL

accession no.

BC011526
NM_145012

shipped in

dry ice

storage temp.

−70°C

Gene Information

Application

Useful for kinase assay and Western blot.

Biochem/physiol Actions

Full-length recombinant human PCTK3 (CDK18) and Cyclin Y (2-end) were co-expressed by Baculovirus in Sf9 insect cells using N-terminal GST tags. PCTK3 or PCTAIRE protein kinase 3 is a member of the cdc2/cdkx subfamily of the ser/thr family of protein kinases. PCTK3 is thought to play a role in terminally differentiated neurons and it contains an N-terminal extension relative to CDC2 (1). PCTK3 is expressed in a variety of human cell lines and tissues (2). Elevation in the protein levels of PCTAIRE 3 in the temporal cortex of Alzheimer′s disease relative to control brains, have been observed. It is proposed that PCTAIRE 3 is a paired helical filaments-associated kinase that modulates tau phosphorylation.

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service