Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0720

Sigma-Aldrich

SBK1, Active

PRECISIO® Kinase, recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

15-25 nmol/min·mg (radiometric)

mol wt

~69 kDa

concentration

0.05-0.1 μg/μL

accession no.

NM_001024401

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SBK1(388228)

Application

Useful for kinase assay.

Biochem/physiol Actions

Full-length recombinant human SBK1 was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. SBKI or SH3 domain binding kinase 1 is a novel serine/threonine protein kinase structurally related to a Xenopus gastrula-specific protein kinase, Pk9.7 which possessing a consensus sequence for an SH3-binding domain from developing rat brain and it play an important role in signal-transduction pathways related to the control of brain development (1). SBK1 may predict a broad cellular function, and its dysregulated in certain cancers suggests an involvement of the protein in the pathogenesis of human cancers (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service