Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0584

Sigma-Aldrich

Human CD46-C, His-Tag

recombinant, expressed in HEK 293 cells

Synonym(s):

membrane co-factor protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

32.8 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CD46(4179)

General description

Human CD46-C, also known as membrane co-factor protein, GenBank Accession No. BC_030594, a.a. 35-285 and 301-328 with C-terminal His-tag, expressed in a HEK293 cell expression system. MW = 35.2 kDa (precursor), 32.8 kDa (mature). Note: this protein is expressed with a 34 amino acid signal sequence that is cleaved after secretion and is heavily glycosylated, resulting in a protein that runs at a higher apparent MW.

Physical form

Aqueous solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service