Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0340

Sigma-Aldrich

Axl active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)

Synonym(s):

AXL receptor tyrosine kinase, JTK11, UFO

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

77 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

activity assay: suitable
inhibition assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AXL(558)

General description

AXL (GenBank Accession No. NM_021913), amino acids 473-894 with N-terminal GST-tag, MW= 77 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme that will transfer 1 nmol phosphate to Tyr substrate per minute at pH 7.4 and 30° C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tateo Sawabu et al.
Molecular carcinogenesis, 46(2), 155-164 (2006-12-23)
Activation of tyrosine kinases is an important factor during cancer development. Axl, one of the receptor tyrosine kinases, binds to the specific ligand growth arrest-specific gene 6 (Gas6), which encodes a vitamin K-dependent gamma-carboxyglutamyl protein. Although many receptor tyrosine kinases
Gaetano Zizzo et al.
Arthritis research & therapy, 15(6), R212-R212 (2013-12-12)
Systemic lupus erythematosus (SLE) is characterized by impaired efferocytosis and aberrant activation of innate immunity. We asked if shedding of MER receptor tyrosine kinase (MerTK) and AXL into soluble (s) ectodomains was related to immunological and clinical aspects of SLE.
M Vouri et al.
Oncogenesis, 5(10), e266-e266 (2016-10-25)
Acquired resistance to conventional and targeted therapies is becoming a major hindrance in cancer management. It is increasingly clear that cancer cells are able to evolve and rewire canonical signalling pathways to their advantage, thus evading cell death and promoting
Peter Vajkoczy et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(15), 5799-5804 (2006-04-06)
Malignant gliomas remain incurable brain tumors because of their diffuse-invasive growth. So far, the genetic and molecular events underlying gliomagenesis are poorly understood. In this study, we have identified the receptor tyrosine kinase Axl as a mediator of glioma growth
Xiao-Han Qu et al.
Oncology letters, 10(3), 1677-1685 (2015-12-02)
Lung cancer has long been one of the most serious types of malignant tumor, and is associated with high incidence and mortality rates. Despite advancements in the comprehensive treatment of the disease, particularly with targeted therapeutic agents, there has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service