SML3658
BMS-470539 dihydrochloride
≥98% (HPLC)
Synonym(s):
(S)-2-Amino-N-((R)-1-(4-butyryl-4-phenylpiperidin-1-yl)-3-(4-methoxyphenyl)-1-oxopropan-2-yl)-3-(1-methyl-1H-imidazol-5-yl)propanamide dihydrochloride, 1-[1-(3-Methyl-L-histidyl-O-methyl-D-tyrosyl)-4-phenyl-4-piperidinyl]-1-Butanone dihydrochloride, BMS 470539 dihydrochloride, BMS470539 dihydrochloride
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
white to beige
solubility
H2O: 2 mg/mL, clear (Warmed)
storage temp.
-10 to -25°C
SMILES string
CCCC(C1(C2=CC=CC=C2)CCN(CC1)C([C@H](NC([C@@H](N)CC3=CN=CN3C)=O)CC4=CC=C(C=C4)OC)=O)=O.[2HCl]
Biochem/physiol Actions
Caution
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service