Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3412

Sigma-Aldrich

Talinolol

≥95% (HPLC)

Synonym(s):

(±)-Talinolol, 02-115, 1-(4-(3-(tert-Butylamino)-2-hydroxypropoxy)phenyl)-3-cyclohexylurea, 1-(4-cyclohexylureidophenoxy)-2-hydroxy-3-tert-butylaminopropane, Cordanum, N-Cyclohexyl-N′-[4-[3-[(1,1-dimethylethyl)amino]-2-hydroxypropoxy]phenyl]urea, rac-Talinolol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H33N3O3
CAS Number:
Molecular Weight:
363.49
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

N(C2CCCCC2)C(=O)Nc1ccc(cc1)OCC(O)CNC(C)(C)C

InChI

1S/C20H33N3O3/c1-20(2,3)21-13-17(24)14-26-18-11-9-16(10-12-18)23-19(25)22-15-7-5-4-6-8-15/h9-12,15,17,21,24H,4-8,13-14H2,1-3H3,(H2,22,23,25)

InChI key

MXFWWQICDIZSOA-UHFFFAOYSA-N

Biochem/physiol Actions

Talinolol is an orally active beta blocker (β blocker) with in vivo antihypertensive efficacy. In addition to its beta-adrenergic receptor (βAR, bAR) antagonist potency (βAR1/2 Ki = 0.24/0.9 μM), talinolol is also reported to exhibit inhibitory side-activity against soluble epoxide hydrolase (sEH IC50 = 2.8 μM).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service