Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2362

Sigma-Aldrich

WRR139

≥98% (HPLC)

Synonym(s):

(S,E)-N-Isopentyl-4-phenyl-2-(3-tosylacrylamido)butanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32N2O4S
CAS Number:
Molecular Weight:
456.60
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

WRR139 is a peptidyl vinyl sulfone that inhibits NGLY1 (N-glycanase 1; PNGase; Peptide-N(4)-(N-acetyl-beta-glucosaminyl)asparagine amidase) de-N-glycosylation activity in cell-free enzymatic assays (IC50 <10 μM; 3.75 rhNGLY1 wtih 1.7 μg S-alkylated RNase B as substrate) and in cultures (IC50 = 5.5 μM in ddVenus reporter K562 cells co-treated with 1 μM proteasome inhibitor carfilzomib). WRR139 disrupts NGLY1-mediated nuclear respiratory factor 1 (NRF1) processing/activation. WRR139 (1 μM) is shown to potentiate the cytotoxicity of proteasome inhibitor carfilzomib (1-100 nM) in multiple myeloma (MM; U266 & H929), T-cell acute lymphoblastic leukemia (T-ALL; Jurkat), and HeLa cultures, but not in NGLY1-knockdown HeLa cells. Note: concentrations below 10 μM is generally recommended for culture treatment to avoid off-target activity against caspases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frederick M Tomlin et al.
ACS central science, 3(11), 1143-1155 (2017-12-05)
Proteasome inhibitors are used to treat blood cancers such as multiple myeloma (MM) and mantle cell lymphoma. The efficacy of these drugs is frequently undermined by acquired resistance. One mechanism of proteasome inhibitor resistance may involve the transcription factor Nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service