Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1725

Sigma-Aldrich

RTC13

≥98% (HPLC)

Synonym(s):

(5Z)-2-Amino-5-[[5-(2-nitrophenyl)-2-furanyl]methylene]-4(5H)-thiazolone, (Z)-2-Amino-5-((5-(2-nitrophenyl)furan-2-yl)methylene)thiazolidin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9N3O4S
CAS Number:
Molecular Weight:
315.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1NC(S/C1=C\C2=CC=C(C3=C([N+]([O-])=O)C=CC=C3)O2)=N

InChI

1S/C14H9N3O4S/c15-14-16-13(18)12(22-14)7-8-5-6-11(21-8)9-3-1-2-4-10(9)17(19)20/h1-7H,(H2,15,16,18)/b12-7+

InChI key

CZZAPCPWFCGOCC-KPKJPENVSA-N

Biochem/physiol Actions

RTC13 is a cell-permeable thiazolidinone derivative that promotes ribosomal read-through activity of premature termination codons (PTCs) and effectively restores full-length protein production from transcripts containing nonsense mutations-generated PTCs, including ataxia-telangiectasia (A-T) mutated in A-T lymphoblastoid cell cultures and dystrophin in mdx Duchenne muscular dystrophy (DMD) mice-derived muscle myotubes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service