Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1459

Sigma-Aldrich

SHA 68

≥98% (HPLC)

Synonym(s):

3-Oxo-1,1-diphenyl-tetrahydro-oxazolo[3,4-a]pyrazine-7-carboxylic acid 4-fluoro-benzylamide, N-[(4-Fluorophenyl)methyl]tetrahydro-3-oxo-1,1-diphenyl-3H-oxazolo[3,4-a]pyrazine-7(1H)-carboxamide, SHA-68

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24FN3O3
CAS Number:
Molecular Weight:
445.49
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

originator

Takeda

storage temp.

2-8°C

InChI

1S/C26H24FN3O3/c27-22-13-11-19(12-14-22)17-28-24(31)29-15-16-30-23(18-29)26(33-25(30)32,20-7-3-1-4-8-20)21-9-5-2-6-10-21/h1-14,23H,15-18H2,(H,28,31)

InChI key

SFRQIPRTNYHJHP-UHFFFAOYSA-N

Biochem/physiol Actions

SHA 68 is a potent and selective neuropeptide S receptor antagonist that exhibits anxiogenic effects.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service