Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1280

Sigma-Aldrich

TT01001

≥98% (HPLC)

Synonym(s):

4-[[[(3,5-Dichlorophenyl)amino]thioxomethyl]amino]-1-piperidinecarboxylic acid ethyl ester, Ethyl4-(3-(3,5-dichlorophenyl)thioureido)piperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19Cl2N3O2S
CAS Number:
Molecular Weight:
376.30
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

TT01001 is potent, orally available the mitochondrial outer membrane protein mitoNEET ligands that binds to mitoNEET without PPARγ activation. TT01001 improves hyperglycemia, hyperlipidemia, and glucose intolerance in mice models of diabetes II. TT01001 exerts anti-diabetic effects without the pioglitazone associated weight gain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service