Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0738

Sigma-Aldrich

RORγ Activator 1b

≥98% (HPLC)

Synonym(s):

N-(4,6-Dimethyl-2-benzothiazolyl)-3-methyl-2-thiophenecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N2OS2
CAS Number:
Molecular Weight:
302.41
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

RORγ Activator 1b binds to the retinoid-related orphan receptor RORγ and induces IL-17 reporter gene expression with an EC50 of 0.1 μM. The compound RORγ Activator 1b enhances differentiation of CD4+ T cells to IL-17-secreting cells with a maximum effect of 220 percent of control at 3 μM.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service